Thermo Scientific Nalgene 2135-8302 Top Works 83B Cap, 1/4" ID (2), 1/Pk
Representative image only.
Thermo Scientific Nalgene 2135-8302 Top Works 83B Cap, 1/4" ID (2), 1/Pk
Thermo Scientific | Mfr: 2135-8302| Item: 1307012
₹167,000.00INR / Each
₹167,000.00INR / Each
Buy It Together
Current Item
Thermo Scientific Nalgene 2135-8302 Top Works 83B Cap, 1/4" ID (2), 1/Pk
Thermo Scientific Nalgene 2135-8302 Top Works 83B Cap, 1/4" ID (2), 1/Pk
1307012
₹167,000.00INR / Each
+ =
Total Price 190100.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Top Works™ closure systems and NALGENE® bottles and carboys create the ideal aseptic fluid transfer system!
  • 6 month warranty
More About this Item
Flexible Top Works™ Closures are available in solid, two-port and three-port versions. Closures are polypropylene (PP). Platinum-cured silicone tubing is fused to a silicone stopper insert which forms a one-piece closure system. Autoclavable and leakproof.
6 Month Warranty
Specifications
  • MaterialPolypropylene(PP)
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size83B
  • Cap ColorWhite
  • AutoclavableYes
  • Tubing ID6.4 mm
  • DescriptionTop Works 83B Cap, 1/4" ID (2), 1/Pk