Thermo Scientific LabTower EDI 15 l/h with EDI cell, storage tank 100 litres and pressure pump 1,5 l/min
Representative image only.
Thermo Scientific LabTower EDI 15 l/h with EDI cell, storage tank 100 litres and pressure pump 1,5 l/min
Thermo Scientific | Mfr: 07.4415| Item: 1595458
Integrated system designed to produce ultra pure water at 18.2MÙcm (0.055µS/cm)
Call For Price
Call For Price
Product Description
Key Features
  • Automatic and economical production of high purity water for daily requirements of 100 to 500 liters/hr
  • 1 year warranty
More About this Item
It combines a pre-treatment cartridge (consisting of a Reverse Osmosis membrane and an activated carbon filter) and an ion exchange cartridge. Purified water can be drawn directly from the system via a dispensing tap (ASTM 1) or from the integrated 100 liter water storage tank (ASTM II)
Microprocessor controlled for automatic operation with permanent monitoring of critical parameters
To ensure the water produced by the Pacific does not deteriorate a recirculation pump periodically passes the purified water back over the Reverse Osmosis membrane. This ensures the stored water is always at the optimal quality and ready to use immediately.
1 Year WarrantyConformite Europeenne
Specifications
  • DescriptionLabTower EDI 15 l/h with EDI cell, storage tank 100 litres and pressure pump 1,5 l/min