Thermo Scientific GenPure UV/TOC ultra pure water system with TOC monitor and UV Intensity
Representative image only.
Thermo Scientific GenPure UV/TOC ultra pure water system with TOC monitor and UV Intensity
Thermo Scientific | Mfr: 08.2206| Item: 1595460
Ultrapure water up to 18. 2 M-cm with UV intensity and TOC monitoring Choose ultrafiltration model 15954-61 for water free of endotoxins and nucleases
Call For Price
Call For Price
Product Description
Key Features
  • Ideal for applications such as trace analysis (HPLC, IC, ICP-MS), cell and tissue culture media, and PCR
  • 1 year warranty
More About this Item
This new generation system offers continuous real-time TOC measurement combined with permanent monitoring of the intensity of the UV lamp. No longer face the risk of false TOC values with decreasing UV intensity! Digital microprocessor control offers fully automatic supervision and control. Real-time clock and code-protected operating system prevent unauthorized changes or resetting.
Sanitization of all parts that contact the water provide optimal microbiological control. Particle-free dispensing through sterile filtration as water is dispensed. Protective bell avoids unintentional hand contact of the outlet.
Complete GLP compliant system password protection and facility to print out data at programmable regular intervals. Complete validation protocols available with traceable calibration certificate.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
1 Year WarrantyConformite Europeenne
Specifications
  • DescriptionGenPure UV/TOC ultra pure water system with TOC monitor and UV Intensity