Thermo Scientific Genesys 4010-000 Replacement Single-Cell Holder
Representative image only.
Thermo Scientific Genesys 4010-000 Replacement Single-Cell Holder
Thermo Scientific | Mfr: 4010-000| Item: 0265094
Optimize performance of your spectrophotometer
₹27,000.00INR / Each
₹27,000.00INR / Each
Product Description
Key Features
  • Choose from a wide range of parts and accessories compatible with the Thermo Scientific™ GENESYS™ spectrophotometer
  • 1 year warranty
More About this Item
Parts and accessories to expand or keep your analysis running smoothly.
Holds one, 10 mm, square cuvet, or one 1/2" (10 mm) diameter test tube.
Available for sale and use in the United States only.
1 Year Warranty
Specifications
  • DescriptionReplacement Single-Cell Holder