Thermo Scientific FL583X1 Pretreatment cartridge, Pore size: 5 µm

Representative image only.

Thermo Scientific FL583X1 Pretreatment cartridge, Pore size: 5 µm

Thermo Scientific | Mfr: FL583X1| Item: 0150213

Fill with your choice of cartridges

₹29,000.00INR / Each

₹29,000.00INR / Each
Product Description

Key Features

  • For half-sized holder system
  • 1 year warranty

More About this Item

Pre-treat to save your fine filters.

1 Year Warranty
Specifications
  • Pore size (µm)5
  • DescriptionPretreatment cartridge, Pore size: 5 µm
  • Warranty1 year
Reviews
Q&A