Thermo Scientific Dual Star Meter Fluoride Kit

Representative image only.

Thermo Scientific Dual Star Meter Fluoride Kit

Thermo Scientific | Mfr: 2115203| Item: 5882513

No manual needed to begin measuring

Call For Price

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

Thermo Scientific Dual Star Meter Fluoride Kit
Thermo Scientific Dual Star Meter Fluoride Kit
5882513

Call For Price
+ =
Total Price 24700.00 ADD 2 ITEMS TO CART
Product Description

Includes

refillable glass-body Orion ROSS Ultra™ pH electrode, pH calibration buffer set and storage solution, ion-selective electrode, ISE standards and ISA solutions set, electrode stand, stainless steel ATC probe, and universal AC power supply.

Key Features

  • Dual inputs allow you to simultaneously measure and display pH, ISE and temperature
  • Simple to operate — prompts to walk you through each meter operation
  • More options for ISE than any other meter
  • 3 year warranty

More About this Item

The Thermo Scientific Orion Dual Star meter provides dual BNC inputs for simultaneous pH and ISE readings. Ion determination is facilitated by additional options for concentration units, optional auto-blank for low-level measurements, built-in Star-stirrer control, and the ability to view and print your calibration graph with slope data. The meter easily guides you through common direct measurements, but also is programmed for a range of Known Addition and Subtraction methods. Autosampler capabilities allow you to take advantage high sample throughput technology to save time and improve measurement quality.


The Dual Star meter uses Smart Stability™ and Averaging™ functions to ensure accurate results. The meter offers a full complement of GLP-related features including 1000 point data logging with time and date stamp, electrode verification, and full calibration data. Generate and store up to 10 password protected methods per channel. Easily download data from the USB port or bidirectional RS-232.


3 Year Warranty
Specifications
  • Meter TypeBenchtop
  • Display typeLCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.1/0.01/0.001
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, USA, and custom
  • CalibrationUp to 6 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1/1
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min ISE0 ppm
  • Max ISE19999 ppm
  • ISE ResolutionUp to 3 digits
  • ISE Accuracy±0.2% mV or 0.05% whichever is greater
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Temperature Resolution0.1
  • Temperature Accuracy±0.1°C
  • Temperature compensationAutomatic or manual
  • InterfaceUSB or RS232
  • Data Logging (points)1000 data points
  • WaterproofYes
  • Width (in)9
  • Height (in)2
  • DescriptionMeter Fluoride Kit
  • Warranty3 year
Reviews
Q&A