Thermo Scientific DO 110 EcoScan Dissolved Oxygen Meter with Galvanic Probe
Representative image only.
Thermo Scientific DO 110 EcoScan Dissolved Oxygen Meter with Galvanic Probe
Thermo Scientific | Mfr: ECDO11001| Item: 9985676
Ideal for testing water quality, wastewater, surface water, groundwater, or drinking water
₹80,500.00INR / Each
₹80,500.00INR / Each
Product Description
Includes
Meter only: Meter, batteries, and Datalog Assist PC software on CD-ROM. Meter and probe: Meter, batteries, DO probe with 10' cable 35640-50, and Datalog Assist PC software on CD-ROM. Meter kit: Meter, batteries, DO probe with 10' cable 35640-50, Datalog
Key Features
  • 3 year warranty
More About this Item
Meter displays dissolved oxygen (DO) readings in mg/L, ppm, or % saturation with simultaneous temperature display. Independent 100% and 0% calibration points provide high accuracy over the entire range. Increase reading accuracy by entering barometric pressure and salinity — meters automatically calculate offset.
Electrode diagnostics are easily completed with the electrode slope, zero offset, and millivolt display options. Download data via optional RS-232 cable (order separately below).
Meter 35640-20 come with 10-ft galvanic DO probe 35640-50.
3 Year Warranty
Specifications
  • Meter TypeAccessory; Probes and Sensors
  • CalibrationSingle or dual point
  • Temperature Accuracy±0.9°F (±0.5°C)
  • Temperature compensationAutomatic or manual
  • InterfaceRS-232
  • Data Logging (points)100 data points
  • Battery4 x AAA batteries (included)
  • Battery life (hr)700
  • DescriptionEcoScan Dissolved Oxygen Meter with Galvanic Probe