Thermo Scientific CIC0001831 Support Rod Clamp and Holder
Representative image only.
Thermo Scientific CIC0001831 Support Rod Clamp and Holder
Thermo Scientific | Mfr: CIC0001831| Item: 0460099
Enhance performance and safety of your stirrers and hotplates
₹15,000.00INR / Each
₹15,000.00INR / Each
Buy It Together
Current Item
Thermo Scientific CIC0001831 Support Rod Clamp and Holder
Thermo Scientific CIC0001831 Support Rod Clamp and Holder
0460099
₹15,000.00INR / Each
+ + +
Traceable Monitoring Hygrometer
2025030
₹6,700.00INR / Each
=
Total Price 455700.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Optimize use of your hotplates and stirrers
  • 3 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories suitable for use with the Thermo Scientific™ Cimarec+™ and Super-Nuova+™ stirrers, hotplates, or hotplate stirrers. Choose accessories compatible with your equipment.
Support rod and clamps are compatible with the integral rod holder built into all Cimarec+ and SuperNuova+ hot plates, stirrers, and stirring hot plates. Ideal for securing flasks, temperature probes, and other apparatus for added safety.
3 Year Warranty
Specifications
  • DescriptionSupport Rod Clamp and Holder