Thermo Scientific CIC0001830 Temperature Probe Support Rod
Representative image only.
Thermo Scientific CIC0001830 Temperature Probe Support Rod
Thermo Scientific | Mfr: CIC0001830| Item: 0460098
Enhance performance and safety of your stirrers and hotplates
₹12,600.00INR / Each
₹12,600.00INR / Each
Buy It Together
Current Item
Thermo Scientific CIC0001830 Temperature Probe Support Rod
Thermo Scientific CIC0001830 Temperature Probe Support Rod
0460098
₹12,600.00INR / Each
+ + +
Traceable Monitoring Hygrometer
2025030
₹6,700.00INR / Each
=
Total Price 453300.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Optimize use of your hotplates and stirrers
  • 3 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories suitable for use with the Thermo Scientific™ Cimarec+™ and Super-Nuova+™ stirrers, hotplates, or hotplate stirrers. Choose accessories compatible with your equipment.
Support rod and clamps are compatible with the integral rod holder built into all Cimarec+ and SuperNuova+ hot plates, stirrers, and stirring hot plates. Ideal for securing flasks, temperature probes, and other apparatus for added safety.
3 Year Warranty
Specifications
  • DescriptionTemperature Probe Support Rod