Thermo Scientific CIC0001471 Replacement Power Cord with US/Canada/Japan Plug; 100-120 VAC, 50/60Hz
Representative image only.
Thermo Scientific CIC0001471 Replacement Power Cord with US/Canada/Japan Plug; 100-120 VAC, 50/60Hz
Thermo Scientific | Mfr: CIC0001471| Item: 0460097
Enhance performance and safety of your stirrers and hotplates
₹11,300.00INR / Each
₹11,300.00INR / Each
Buy It Together
Current Item
Thermo Scientific CIC0001471 Replacement Power Cord with US/Canada/Japan Plug; 100-120 VAC, 50/60Hz
Thermo Scientific CIC0001471 Replacement Power Cord with US/Canada/Japan Plug; 100-120 VAC, 50/60Hz
0460097
₹11,300.00INR / Each
+ + +
Traceable Monitoring Hygrometer
2025030
₹6,700.00INR / Each
=
Total Price 452000.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Optimize use of your hotplates and stirrers
  • 3 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories suitable for use with the Thermo Scientific™ Cimarec+™ and Super-Nuova+™ stirrers, hotplates, or hotplate stirrers. Choose accessories compatible with your equipment.
Power cord includes a connector for installation in the hot plate, stirrer, or stirring hot plate and a plug. Cord is suitable for 100 to 120 VAC, 50/60 Hz power for use in USA, Canada, and Japan and is three-pronged.
3 Year Warranty
Specifications
  • Power (VAC)100 to 120
  • DescriptionPower Cord; 100-120V US/JP