Thermo Scientific CIC0001447 External Temperature Probe, Stainless Steel, PT100, 0-400°C
Representative image only.
Thermo Scientific CIC0001447 External Temperature Probe, Stainless Steel, PT100, 0-400°C
Thermo Scientific | Mfr: CIC0001447| Item: 0460091
Enhance performance and safety of your stirrers and hotplates
₹30,500.00INR / Each
₹30,500.00INR / Each
Buy It Together
Current Item
Thermo Scientific CIC0001447 External Temperature Probe, Stainless Steel, PT100, 0-400°C
Thermo Scientific CIC0001447 External Temperature Probe, Stainless Steel, PT100, 0-400°C
0460091
₹30,500.00INR / Each
+
Traceable Monitoring Hygrometer
2025030
₹6,700.00INR / Each
+ + =
Total Price 471200.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Optimize use of your hotplates and stirrers
  • Ideal for use with Cimarec and Supernuova hotplates and hotplate stirrers
  • 3 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories suitable for use with the Thermo Scientific™ Cimarec+™ and Super-Nuova+™ stirrers, hotplates, or hotplate stirrers. Choose accessories compatible with your equipment.
External temperature probe works with both Cimarec+ and SuperNuova+ hot plates, stirrers, and stirring hot plates. Standard stainless steel is suitable for most laboratory environments. Maximum temperature is 752°F (400°C).
3 Year Warranty
Specifications
  • Min Temperature (° F)32
  • Max Temperature (° F)752
  • Min Temperature (DegreeC)0
  • Max Temperature (° C)400
  • DescriptionExternal Probe, Stainlss Steel, PT100