Thermo Scientific CIC0001445 Top Plate Shield, Transparent, 7x7"

Representative image only.

Thermo Scientific CIC0001445 Top Plate Shield, Transparent, 7x7"

Thermo Scientific | Mfr: CIC0001445| Item: 0460089

Enhance performance and safety of your stirrers and hotplates

₹9,100.00INR / Each

₹9,100.00INR / Each

Buy It Together

Current Item

Thermo Scientific CIC0001445 Top Plate Shield, Transparent, 7x7"
Thermo Scientific CIC0001445 Top Plate Shield, Transparent, 7x7"
0460089

₹9,100.00INR / Each
+ + +
Traceable Monitoring Hygrometer
2025030

₹6,700.00INR / Each
=
Total Price 449800.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Optimize use of your hotplates and stirrers
  • 3 year warranty

More About this Item

Choose from a wide range of replacement parts and accessories suitable for use with the Thermo Scientific™ Cimarec+™ and Super-Nuova+™ stirrers, hotplates, or hotplate stirrers. Choose accessories compatible with your equipment.

Transparent, removable shield/splash guard protects samples and users. Universal design fits all Cimarec+ and SuperNuova+ hot plates, stirrers, and stirring hot plates.

3 Year Warranty
Specifications
  • Top Plate Length (in)7
  • Top Plate Width (in)7
  • Top Plate Length (cm)17.78
  • Top Plate Width (cm)17.78
  • DescriptionTop Plate Shield; 7x7"
  • Warranty3 year
Reviews
Q&A