Thermo Scientific Barnstead D8905 Disposable Ion Exchange Cartridge, cation removal
Representative image only.
Thermo Scientific Barnstead D8905 Disposable Ion Exchange Cartridge, cation removal
Thermo Scientific | Mfr: D8905| Item: 9926006
Customize your system with application-specific water purification cartridges
₹38,100.00INR / Each
₹38,100.00INR / Each
Buy It Together
Current Item
Thermo Scientific Barnstead D8905 Disposable Ion Exchange Cartridge, cation removal
Thermo Scientific Barnstead D8905 Disposable Ion Exchange Cartridge, cation removal
9926006
₹38,100.00INR / Each
+ + + =
Total Price 118500.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Disposable cartridges are ideal for use in nonpressurized applications
  • Inert polypropylene cartridge magerial prevents contamination
  • 2 year warranty
More About this Item
Order a cartridge holder (sold separately) to support your cartridges.
Color-changing resin indicates when cartridge is exhausted. Cannot be used with alcohol-containing fluids.
Two year warranty
Specifications
  • MediaResin
  • Capacity (grains)3000 as Sodium Chloride
  • Connection3/8" hose barb at each end
  • FunctionCation exchange
  • Max Flow Rate (LPH)1.25
  • Diameter (in)3.4375
  • DescriptionDisposable Ion Exchange Cartridge, cation removal