Thermo Scientific Barnstead D8901 Ion Exchange Cartridge; Two-Bed
Representative image only.
Thermo Scientific Barnstead D8901 Ion Exchange Cartridge; Two-Bed
Thermo Scientific | Mfr: D8901| Item: 9926002
Customize your system with application-specific water purification cartridges
₹50,400.00INR / Each
₹50,400.00INR / Each
Buy It Together
Current Item
Thermo Scientific Barnstead D8901 Ion Exchange Cartridge; Two-Bed
Thermo Scientific Barnstead D8901 Ion Exchange Cartridge; Two-Bed
9926002
₹50,400.00INR / Each
+ + + =
Total Price 130800.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Disposable cartridges are ideal for use in nonpressurized applications
  • Inert polypropylene cartridge magerial prevents contamination
  • 1 year warranty
More About this Item
Order a cartridge holder (sold separately) to support your cartridges.
Color-changing resin indicates when cartridge is exhausted. Cannot be used with alcohol-containing fluids.
1 Year Warranty
Specifications
  • MediaResin
  • Capacity (grains)1725 as Sodium Chloride
  • Connection3/8" hose barb at each end
  • FunctionHigh capacity, lower purity
  • Max Flow Rate (LPH)1.25
  • Diameter (in)3.4375
  • DescriptionIon Exchange Cartridge; Two-Bed