Thermo Scientific Barnstead D8822 Ion Exchange Cartridge; Half-Size, Combo
Representative image only.
Thermo Scientific Barnstead D8822 Ion Exchange Cartridge; Half-Size, Combo
Thermo Scientific | Mfr: D8822| Item: 9926000
Customize your system with application-specific water purification cartridges
₹51,000.00INR / Each
₹51,000.00INR / Each
Buy It Together
Current Item
Thermo Scientific Barnstead D8822 Ion Exchange Cartridge; Half-Size, Combo
Thermo Scientific Barnstead D8822 Ion Exchange Cartridge; Half-Size, Combo
9926000
₹51,000.00INR / Each
+ + + =
Total Price 131400.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Disposable cartridges are ideal for use in nonpressurized applications
  • Inert polypropylene cartridge magerial prevents contamination
  • 1 year warranty
More About this Item
Order a cartridge holder (sold separately) to support your cartridges.
1 Year Warranty
Specifications
  • MediaCombination resin
  • Capacity (grains)190 as Sodium Chloride
  • Connection3/8" hose barb at each end
  • FunctionIon exchange, oxygen removal
  • Max Flow Rate (LPH)1.25
  • Diameter (in)3.4375
  • DescriptionIon Exchange Cartridge; Half-Size, Combo