Thermo Scientific Barnstead D5029 Type I cartridge kit, three module
Representative image only.
Thermo Scientific Barnstead D5029 Type I cartridge kit, three module
Thermo Scientific | Mfr: D5029| Item: 9925811
Select pretreatment, deionization, or organic-free cartridges, depending upon
your application
₹169,000.00INR / Each
₹169,000.00INR / Each
Product Description
Key Features
  • For use with E-Pure systems
  • Produce MΩ-cm resistivity, Type I reagentgrade water
More About this Item
Cartridge kits include all cartridges needed to produce 18.3 MΩ-cm resistivity, Type I reagentgrade water. Kits that include organic-free cartridges produce water that has less than 10 ppb
organic carbon. Kits for Nanopure Infinity and Nanopure Dlamond systems include final filter; order final filter for quad-style Nanopure and E-Pure systems separately.
Kit contains one 01505-22 and two 01505-29.
Specifications
  • DescriptionType I cartridge kit, three module