Thermo Scientific Barnstead D50230 pretreatment cartridge
Representative image only.
Thermo Scientific Barnstead D50230 pretreatment cartridge
Thermo Scientific | Mfr: D5023| Item: 9925822
Select pretreatment, deionization, or organic-free cartridges, depending upon
your application
₹274,000.00INR / Each
₹274,000.00INR / Each
Product Description
Key Features
  • For use with E-Pure systems
  • Produce MΩ-cm resistivity, Type I reagentgrade water
  • 1 year warranty
More About this Item
Cartridge kits include all cartridges needed to produce 18.3 MΩ-cm resistivity, Type I reagentgrade water. Kits that include organic-free cartridges produce water that has less than 10 ppb
organic carbon. Kits for Nanopure Infinity and Nanopure Dlamond systems include final filter; order final filter for quad-style Nanopure and E-Pure systems separately.
Kit contains one 99257-00, one 01505-24, one 01505-29, and one 01506-05.
1 Year Warranty
Specifications
  • DescriptionType I cartridge kit, organic free, four module