Thermo Scientific Barnstead D50220 Ion Exchange Cartridge; Half-Size, Mixed-Bed
Representative image only.
Thermo Scientific Barnstead D50220 Ion Exchange Cartridge; Half-Size, Mixed-Bed
Thermo Scientific | Mfr: D50220| Item: 9926001
Customize your system with application-specific water purification cartridges
₹33,600.00INR / Each
₹33,600.00INR / Each
Buy It Together
Current Item
Thermo Scientific Barnstead D50220 Ion Exchange Cartridge; Half-Size, Mixed-Bed
Thermo Scientific Barnstead D50220 Ion Exchange Cartridge; Half-Size, Mixed-Bed
9926001
₹33,600.00INR / Each
+ + + =
Total Price 114000.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Disposable cartridges are ideal for use in nonpressurized applications
  • Inert polypropylene cartridge magerial prevents contamination
  • 2 year warranty
More About this Item
Order a cartridge holder (sold separately) to support your cartridges.
Two year warranty
Specifications
  • MediaResin
  • Capacity (grains)430 as Sodium Chloride
  • Connection3/8" hose barb at each end
  • FunctionUltrapure ion exchange
  • Max Flow Rate (LPH)1.25
  • Diameter (in)3.4375
  • DescriptionIon Exchange Cartridge; Half-Size, Mixed-Bed