Thermo Scientific Barnstead D502124 Ultra-Low Organics DI Feed
Representative image only.
Thermo Scientific Barnstead D502124 Ultra-Low Organics DI Feed
Thermo Scientific | Mfr: D502124 - BARNSTEAD| Item: 9926180
Call For Price
Call For Price
Product Description
Key Features
  • 1 year warranty
More About this Item
99261-80 has a 430 grain capacity.
99261-82 has a 465 grain capacity.
99261-84 has a 540 grain capacity.
99261-86 has a 575 grain capacity.
1 Year Warranty
Specifications
  • Compatible withEASYpure® II System
  • ApplicationsUltra-low organics and high resistivity
  • DescriptionUltra-Low Organics DI Feed