Thermo Scientific Barnstead D4802 Type I Cartridge Kit, organic free, quad Nanopure
Representative image only.
Thermo Scientific Barnstead D4802 Type I Cartridge Kit, organic free, quad Nanopure
Thermo Scientific | Mfr: D4802| Item: 9925802
Keep your system operating precisely
₹331,000.00INR / Each
₹331,000.00INR / Each
Product Description
Key Features
  • Use with quad-style Nanopure® systems
  • Kits include all cartridges needed to produce 18.3 MΩ-cm resistivity, Type I reagent grade water
  • 1 year warranty
More About this Item
Kit contains one 99257-00, 01505-24, and one 01506-05.
1 Year Warranty
Specifications
  • DescriptionType I Cartridge Kit, organic free, quad Nanopure