Thermo Scientific Barnstead D0836 Pretreatment Cartridge for E-Pure, Nanopure
Representative image only.
Thermo Scientific Barnstead D0836 Pretreatment Cartridge for E-Pure, Nanopure
Thermo Scientific | Mfr: D0836| Item: 9925700
₹67,300.00INR / Each
₹67,300.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Maintain water quality with these replacement cartridges.
1 Year Warranty
Specifications
  • CapacityAs determined by feed water quality
  • Capacity (Liters)7570
  • Capacity (Gal)2000
  • Compatible withAll NANOpure™, E-pure™ systems
  • FunctionRemoves colloids and bacteria; maximizes deionization resin life
  • DescriptionPretreatment Cartridge for E-Pure, Nanopure