Thermo Scientific Barnstead D0835 Pretreatment Cartridge for B-Pure, E-Pure, Nanopure
Representative image only.
Thermo Scientific Barnstead D0835 Pretreatment Cartridge for B-Pure, E-Pure, Nanopure
Thermo Scientific | Mfr: D0835| Item: 0150522
₹50,100.00INR / Each
₹50,100.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Maintain water quality with these replacement cartridges.
1 Year Warranty
Specifications
  • CapacityAs determined by feed water quality
  • Capacity (Liters)18925
  • Capacity (Gal)5000
  • Compatible withAll NANOpure™ E-pure™, B-pure™ systems (except Nanopure Infinity)
  • FunctionRemoves colloids, chlorine, and organics; use when nonionized solids and/or colloids are present
  • DescriptionPretreatment Cartridge for B-Pure, E-Pure, Nanopure