Thermo Scientific Barnstead D0832 Pretreatment Cartridge f/ B-Pure & Bantam, 785 grain
Representative image only.
Thermo Scientific Barnstead D0832 Pretreatment Cartridge f/ B-Pure & Bantam, 785 grain
Thermo Scientific | Mfr: D0832| Item: 0153515
₹46,700.00INR / Each
₹46,700.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Maintain water quality with these replacement cartridges.
1 Year Warranty
Specifications
  • Capacity (grains)785 as Calcium Carbonate
  • Compatible withB-pure™ systems, Bantam
  • FunctionRemoves organics and chlorine
  • DescriptionPretreatment Cartridge f/ B-Pure & Bantam, 785 grain