Thermo Scientific Barnstead D0813 Pretreatment Cartridge f/ B-Pure & Bantam, 5000 gal
Representative image only.
Thermo Scientific Barnstead D0813 Pretreatment Cartridge f/ B-Pure & Bantam, 5000 gal
Thermo Scientific | Mfr: D0813| Item: 0150528
₹42,300.00INR / Each
₹42,300.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Maintain water quality with these replacement cartridges.
1 Year Warranty
Specifications
  • Capacity (Liters)18925
  • Capacity (Gal)5000
  • Compatible withB-pure™ systems, Bantam
  • FunctionRemoves organics and chlorine; maximizes deionization resin life
  • DescriptionPretreatment Cartridge f/ B-Pure & Bantam, 5000 gal