Thermo Scientific Barnstead 50132132 Pacific TII Water Purification Systems 12 l/hr UV

Representative image only.

Thermo Scientific Barnstead 50132132 Pacific TII Water Purification Systems 12 l/hr UV

Thermo Scientific | Mfr: 50132132| Item: 9929026

Multiple capacities available to support your daily requirements

Call For Price

Call For Price

Buy It Together

Current Item

Thermo Scientific Barnstead 50132132 Pacific TII Water Purification Systems 12 l/hr UV
Thermo Scientific Barnstead 50132132 Pacific TII Water Purification Systems 12 l/hr UV
9929026

Call For Price
+
Cole-Parmer WS-200-4 Water Still, 240 VAC
9929500

₹285,000.00INR / Each
+
Cole-Parmer WS-100-4-120 Water Still, 4 L/hr; 120 VAC, 60 Hz
9929502

₹202,000.00INR / Each
+ =
Total Price 784000.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Includes

high purity water cartridge, RO membrane, pressure regulator, built-in wall bracket, and power cord. NOTE: Order required reservoir separately.

Key Features

  • Microprocessor controlled for automatic operation with continuous monitoring of all critical parameters
  • Reservoir fill level (%) automatically shown on display
  • 1 year warranty

More About this Item

Systems deliver up to 240 L/day of Type II pure water from a tap feed water supply. Ideal for routine lab work such as reagent preparation, media preparation, and glassware rinsing. High performance reverse osmosis membrane removes approximately 98% of inorganic ions and 99% of all dissolved organic substances, as well as micoorganisms and particles.

Space-saving design allows benchtop operation or wall mounting. Large, illuminated four-line display is easy to read and clearly shows operating mode status (production, standby, disinfection), reservoir volume (%), and temperature (°C). Display can be tilted for optimal viewing.

Use the RS-232 interface for safe documentation of all measured values and faults with date and time in compliance with GLP guidelines; order external printer separately. Optional UV lamp (254 nm) acts as a germicide, preventing biological contamination.

Opaque polyethylene reservoirs (required; order separately) allow complete draining and efficient cleaning and disinfection. Reservoirs are available with an optional recirculation pump which protects high purity water from bacterial growth during standstill. The 30- and 60-L units can be used with the optional wall-mounting bracket for further space-saving capabilities.


UV lamp (254 nm) acts as a germicide, preventing biological contamination.


1 Year WarrantyConformite Europeenne
Specifications
  • ASTM Water GradeType II
  • Max Production (L/hour)12
  • Max Flow Rate (L/hour)12
  • Inlet PortR 3/4"
  • With UV LampYes
  • Wavelength (nm)254
  • Feed Water Requirements (Source)tap water, softened or hardness stabilized
  • Feed Water Requirements (Conductivity)<1500 µS/cm
  • Feed Water Requirements (pH range)4 to 11
  • Feed Water Requirements (pressure)1.4 to 87 psi (0.1 to 6 bar)
  • Type II Water (Resistivity)15 to 10 MΩ-cm
  • Feed Water Requirements (Temperature)2-35⁰C
  • Type II Water (TOC)<30 ppb
  • Type II Water (Conductivity)0.067 to 0.1 µS-cm
  • DescriptionPacific TII Water Purification Systems 12 l/hr UV
  • Warranty1 year
Reviews
Q&A