Thermo Scientific A2 Non-recirculating cell, 20 x 25 cm gel size

Representative image only.

Thermo Scientific A2 Non-recirculating cell, 20 x 25 cm gel size

Thermo Scientific | Mfr: A2-BP| Item: 2855320

Longer path length for more detailed DNA/RNA analysis

₹348,000.00INR / Each

₹348,000.00INR / Each

Buy It Together

Current Item

Thermo Scientific A2 Non-recirculating cell, 20 x 25 cm gel size
Thermo Scientific A2 Non-recirculating cell, 20 x 25 cm gel size
2855320

₹348,000.00INR / Each
+ =
Total Price 372600.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Base functions as a built-in three-point leveling platform for flat, uniformly thick gels
  • Pour and run gels right in the gasketed gel tray — no taping or sealing needed
  • UV transparent gel tray lets you view DNA bands after performing runs
  • 3 year warranty

More About this Item

What's included: buffer chamber, UV transparent gel tray with gasket end gates, safety lid with attached power leads, and built-in leveling base. All combs are 1.5-mm thick.
Item 28553-20 also includes one each 16-, 24-, and 36-well combs.

3 Year Warranty
Specifications
  • Buffer Solution Required (mL)2300
  • Gel Thickness (mm)1.5
  • Gel Length (cm)25
  • Gel Width (cm)20
  • Cell Width (in)11
  • Cell Height (in)4
  • Cell Depth (in)14.75
  • Cell Width (cm)27.94
  • Cell Height (cm)10.16
  • Cell Depth (cm)37.47
  • DescriptionNon-recirculating cell, 20 x 25 cm gel size
  • Warranty3 year
Reviews
Q&A