Thermo Scientific 977011 Acid adjuster (Ionic Strength and pH Adjusters for Ion-Selective Electrodes)
Representative image only.
Thermo Scientific 977011 Acid adjuster (Ionic Strength and pH Adjusters for Ion-Selective Electrodes)
Thermo Scientific | Mfr: 977011| Item: 0573036
Use these ionic strength and pH adjustors for optimal performance of your ISE
Call For Price
Call For Price
Buy It Together
Current Item
Thermo Scientific 977011 Acid adjuster (Ionic Strength and pH Adjusters for Ion-Selective Electrodes)
Thermo Scientific 977011 Acid adjuster (Ionic Strength and pH Adjusters for Ion-Selective Electrodes)
0573036
Call For Price
+ =
Total Price 21500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Calibrate your instrument and help perform addition and subtraction operations
More About this Item
Combine with ionic strength and pH adjusters to eliminate interferences from other ion species and adjust standards and samples to an optimal pH range. Standardizing and filling solutions as well as ion-selective electrodes are sold separately.
Specifications
  • Ion-Selective ElectrodeChlorine
  • DescriptionAcid adjuster (Ionic Strength and pH Adjusters for Ion-Selective Electrodes)