Thermo Scientific 977010 Iodide Reagent for Chlorine ISE Electrode, 5 x 50 mL
Representative image only.
Thermo Scientific 977010 Iodide Reagent for Chlorine ISE Electrode, 5 x 50 mL
Thermo Scientific | Mfr: 977010| Item: 0573053
Use these ionic strength and pH adjustors for optimal performance of your ISE
₹54,500.00INR / Carton of 5
₹54,500.00INR / Carton of 5
Buy It Together
Current Item
Thermo Scientific 977010 Iodide Reagent for Chlorine ISE Electrode, 5 x 50 mL
Thermo Scientific 977010 Iodide Reagent for Chlorine ISE Electrode, 5 x 50 mL
0573053
₹54,500.00INR / Carton of 5
+ =
Total Price 76000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Calibrate your instrument and help perform addition and subtraction operations
More About this Item
Combine with ionic strength and pH adjusters to eliminate interferences from other ion species and adjust standards and samples to an optimal pH range. Standardizing and filling solutions as well as ion-selective electrodes are sold separately.
Specifications
  • Ion-Selective ElectrodeChlorine
  • DescriptionIodide Reagent for Chlorine ISE Electrode, 5 x 50 mL