Thermo Scientific 967901 ORP Standard, one 475 mL bottle

Representative image only.

Thermo Scientific 967901 ORP Standard, one 475 mL bottle

Thermo Scientific | Mfr: 967901| Item: 0573055

No hazardous chemicals or organics

₹33,100.00INR / Each

₹33,100.00INR / Each

Buy It Together

Current Item

Thermo Scientific 967901 ORP Standard, one 475 mL bottle
Thermo Scientific 967901 ORP Standard, one 475 mL bottle
0573055

₹33,100.00INR / Each
+
LabChem ORP Standard, 200 mV; 500 mL
8002998

Call For Price
=
Total Price 33100.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Use as relative mV standard and calibrate offset of sample
  • Redox potential values are 215 mV for Ag/AgCl reference electrodes and 420 mV for standard hydrogen reference electrodes

More About this Item

These stable, ready-to-use standards require no mixing and contain no organics or hazardous chemicals — use straight out of the bottle. Choose from two convenient package sizes for use in the lab or field.

Specifications
  • Volume (mL)475
  • DescriptionORP Standard, one 475 mL bottle
Reviews
Q&A