Thermo Scientific 951202 Filling solution f/ammonia ISE electrode
Representative image only.
Thermo Scientific 951202 Filling solution f/ammonia ISE electrode
Thermo Scientific | Mfr: 951202| Item: 0573045
Filling solutions for use with pH electrodes and reference electrodes
₹21,100.00INR / Each
₹21,100.00INR / Each
Buy It Together
Current Item
Thermo Scientific 951202 Filling solution f/ammonia ISE electrode
Thermo Scientific 951202 Filling solution f/ammonia ISE electrode
0573045
₹21,100.00INR / Each
+ +
Thermo Scientific 810001 Storage Soluton, 475 mL bottle
0573060
₹18,000.00INR / Each
+ =
Total Price 136110.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Solutions work best when combined with ionic strength and pH adjusters, which eliminate interferences from other ion species
More About this Item
All filling solutions for reference electrodes come in cartons of one 60-mL or five 50-mL bottles.
Specifications
  • Ion-Selective ElectrodeAmmonium
  • DescriptionFilling solution f/ammonia ISE electrode