Thermo Scientific 951011 Ionic Strength and pH Adjustors, Alkaline Reagent, 10 ppm; 475 mL
Representative image only.
Thermo Scientific 951011 Ionic Strength and pH Adjustors, Alkaline Reagent, 10 ppm; 475 mL
Thermo Scientific | Mfr: 951011| Item: 0573044
Use these ionic strength and pH adjustors for optimal performance of your ISE
Call For Price
Call For Price
Buy It Together
Current Item
Thermo Scientific 951011 Ionic Strength and pH Adjustors, Alkaline Reagent, 10 ppm; 475 mL
Thermo Scientific 951011 Ionic Strength and pH Adjustors, Alkaline Reagent, 10 ppm; 475 mL
0573044
Call For Price
+ =
Total Price 21500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Calibrate your instrument and help perform addition and subtraction operations
More About this Item
Combine with ionic strength and pH adjusters to eliminate interferences from other ion species and adjust standards and samples to an optimal pH range. Standardizing and filling solutions as well as ion-selective electrodes are sold separately.
Specifications
  • Ion-Selective ElectrodeCyanide
  • DescriptionIonic strength and pH adjustor; Alkaline reagent