Thermo Scientific 945306 Standardizing Solution, 0.1 M NaI
Representative image only.
Thermo Scientific 945306 Standardizing Solution, 0.1 M NaI
Thermo Scientific | Mfr: 945306| Item: 0573023
Standardizing solutions calibrate your instrument
₹60,600.00INR / Each
₹60,600.00INR / Each
Product Description
Key Features
  • Help perform addition and subtraction operations
More About this Item
Solutions work best when combined with ionic strength and pH adjustors, which eliminate interferences from other ion species and adjust standards and samples to an optimal pH range. All standardizing solutions and ionic strength adjustors come in 475-ml bottles except model 05730-15, (1 gal), and model 05730-53, (five 50-ml bottles). All filling solutions for reference electrodes come in cartons of one or five 50-ml bottles.
Specifications
  • Ion-Selective ElectrodeSodium Iodide
  • DescriptionStandardizing Solution, 0.1 M NaI