Thermo Scientific 932011 Ionic strength and pH adjustors, ISA 4 M KCl
Representative image only.
Thermo Scientific 932011 Ionic strength and pH adjustors, ISA 4 M KCl
Thermo Scientific | Mfr: 932011| Item: 0573010
Use these ionic strength and pH adjustors for optimal performance of your ISE
₹31,000.00INR / Each
₹31,000.00INR / Each
Buy It Together
Current Item
Thermo Scientific 932011 Ionic strength and pH adjustors, ISA 4 M KCl
Thermo Scientific 932011 Ionic strength and pH adjustors, ISA 4 M KCl
0573010
₹31,000.00INR / Each
+ =
Total Price 52500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Calibrate your instrument and help perform addition and subtraction operations
  • 3 Month Product / 12 Month Parts and Labor Warranty
More About this Item
Combine with ionic strength and pH adjusters to eliminate interferences from other ion species and adjust standards and samples to an optimal pH range. Standardizing and filling solutions as well as ion-selective electrodes are sold separately.
Specifications
  • Ion-Selective ElectrodeCalcium
  • DescriptionIonic strength and pH adjustors, ISA 4 M KCl
  • Warranty3 Month Product / 12 Month Parts and Labor