Thermo Scientific 930711 Ammonium Sulfate Ionic Strength Adjustor, 10 ppm; 475 mL
Representative image only.
Thermo Scientific 930711 Ammonium Sulfate Ionic Strength Adjustor, 10 ppm; 475 mL
Thermo Scientific | Mfr: 930711| Item: 0573004
Use these ionic strength and pH adjustors for optimal performance of your ISE
₹270.00INR / Each
₹270.00INR / Each
Buy It Together
Current Item
Thermo Scientific 930711 Ammonium Sulfate Ionic Strength Adjustor, 10 ppm; 475 mL
Thermo Scientific 930711 Ammonium Sulfate Ionic Strength Adjustor, 10 ppm; 475 mL
0573004
₹270.00INR / Each
+ =
Total Price 21770.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Calibrate your instrument and help perform addition and subtraction operations
More About this Item
Combine with ionic strength and pH adjusters to eliminate interferences from other ion species and adjust standards and samples to an optimal pH range. Standardizing and filling solutions as well as ion-selective electrodes are sold separately.
Specifications
  • Ion-Selective ElectrodeNitrate
  • DescriptionAmmonium sulfate adjuster 2m (Ionic Strength/pH Adjusters)