Thermo Scientific 912600 pH Electode, Sealed/SJ/Epoxy/Long-Neck; BNC
Representative image only.
Thermo Scientific 912600 pH Electode, Sealed/SJ/Epoxy/Long-Neck; BNC
Thermo Scientific | Mfr: 912600| Item: 0571151
Value-priced, durable and low maintenance
₹63,300.00INR / Each
₹63,300.00INR / Each
Product Description
Key Features
  • Low cost epoxy body
  • 3 Month Product / 6 Month Parts and Labor Warranty
More About this Item
Measure the pH of virtually any sample. Sealed, gel-filled design means there is little electrode maintenance to perform. Single-junction reference means electrodes are suited to most general applications.
Specifications
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Reference JunctionWick
  • Reference ElectrolyteAg/AgCl
  • Min pH (pH)0
  • Max pH (pH)12
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ConnectorBNC
  • Cable Length (m)1
  • DescriptionpH Electode, Sealed/SJ/Epoxy/Long-Neck; BNC
  • Warranty3 Month Product / 6 Month Parts and Labor