Thermo Scientific 9104APWP AquaPro pH Electode, Sealed/DJ/Glass; WP BNC
Representative image only.
Thermo Scientific 9104APWP AquaPro pH Electode, Sealed/DJ/Glass; WP BNC
Thermo Scientific | Mfr: 9104APWP| Item: 0571133
Ideal for environmental and field testing of water and wastewater samples
₹148,000.00INR / Each
₹148,000.00INR / Each
Product Description
Key Features
  • Low maintenance polymer-filled double-junction electrodes with an extended life, fast response and clog-resistant junction.
  • 1 year warranty
More About this Item
Thermo Scientific Orion AquaPro Professional pH Electrodes are suited for a wide range of applications. Sealed, polymer-filled design provides for little required maintenance. Double-junction extends life and makes electrodes compatible with a wide range of samples including TRIS buffer. All electrodes include a 3.3-ft (1-m) cable.
1 Year Warranty
Specifications
  • ApplicationsSealed
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • Min pH (pH)0
  • Max pH (pH)14
  • Accuracy±0.02 pH
  • Min Temperature (° C)0
  • Max Temperature (° C)60
  • ATC ConnectorNone
  • ConnectorBNC (waterproof when used with Thermo Scientific Orion Star Series meters)
  • Diameter (mm)12
  • Length (mm)120
  • Cable Length (m)1
  • DescriptionAquaPro pH Electode, Sealed/DJ/Glass; WP BNC