Thermo Scientific 910199 Standard All-in-One pH Buffer Kit
Representative image only.
Thermo Scientific 910199 Standard All-in-One pH Buffer Kit
Thermo Scientific | Mfr: 910199| Item: 5535068
Common calibration values are color-coded for ease of use
₹23,000.00INR / Each
₹23,000.00INR / Each
Buy It Together
Current Item
Thermo Scientific 910199 Standard All-in-One pH Buffer Kit
Thermo Scientific 910199 Standard All-in-One pH Buffer Kit
5535068
₹23,000.00INR / Each
+ =
Total Price 26600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
475 mL bottles of each 4.01, 7.00, and 10.01 pH buffers, as well as pH electrode storage solution, and an electrode storage bottle.
Key Features
  • Traceable to NIST standards
More About this Item
Each 475-mL (pint) buffer bottle comes color-coded for easy identification and packaged with buffer/temperature tables for accurate calibrations. Buffers are standardized against NIST-traceable pH references for superior accuracy.
Specifications
  • Buffer Kit Values4.01, 7.00, 10.01 pH buffers, storage solution, and storage bottle
  • Volume (mL)475
  • ColorColored
  • DescriptionStandard All-in-One pH Buffer Kit