Thermo Scientific 910104 pH Buffer Solution, 4.01, Pink; 475 mL Bottle

Representative image only.

Thermo Scientific 910104 pH Buffer Solution, 4.01, Pink; 475 mL Bottle

Thermo Scientific | Mfr: 910104| Item: 5535042

Common calibration values are color-coded for ease of use

₹4,000.00INR / Each

₹4,000.00INR / Each

Buy It Together

Current Item

Thermo Scientific 910104 pH Buffer Solution, 4.01, Pink; 475 mL Bottle
Thermo Scientific 910104 pH Buffer Solution, 4.01, Pink; 475 mL Bottle
5535042

₹4,000.00INR / Each
+ =
Total Price 7600.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Traceable to NIST standards

More About this Item

Each 475-mL (pint) buffer bottle comes color-coded for easy identification and packaged with buffer/temperature tables for accurate calibrations. Buffers are standardized against NIST-traceable pH references for superior accuracy.

Specifications
  • Buffer Value4.01
  • Buffer Kit Values4.01
  • Volume (mL)475
  • ColorPink
  • DescriptionpH Buffer Solution, 4.01, Pink; 475 mL Bottle
Documents & Certificates
Reviews
Q&A