Thermo Scientific 90112 Pipettor Electronic 8-Channel 5-100 uL
Representative image only.
Thermo Scientific 90112 Pipettor Electronic 8-Channel 5-100 uL
Thermo Scientific | Mfr: 90112| Item: 9470265
Pipette, dispense, dilute, and mix with one instrument
Call For Price
for an additional
₹14,500.00INR / Each
for an additional
₹25,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Thermo Scientific 90112 Pipettor Electronic 8-Channel 5-100 uL
Thermo Scientific 90112 Pipettor Electronic 8-Channel 5-100 uL
9470265
Call For Price
+ =
Total Price 14500.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Lightweight ergonomic design
  • Feather-touch piston action and whole-hand tip ejector helps prevent Work Related Upper Limb Disorders (WRULDs)
More About this Item
Features smooth, computerized drive system. Select from five aspiration and five dispensing settings for use with different sample viscosities. Ejector collar and tip come off for easy cleaning. Pipettors operate on factory-installed rechargeable NiMH battery.
Multi-channel models feature ergonomic rotating heads for optimum dispensing. Individual pistons allow for easy repair. Tip cone design ensures easy
tip mounting from trays.
Specifications
  • SterileNo
  • Max Volume (µL)100
  • Min Volume (µL)5
  • Min Volume (mL)0.005
  • Max Volume (mL)0.1
  • Min Dispensing (mL)0.005
  • Max Dispensing (mL)0.1
  • DescriptionPipettor Electronic 8-Channel 5-100 uL