Thermo Scientific 900062 Optimum Results B ISE Fill Solution; Cl/CN/Pb/ Ag/S
Representative image only.
Thermo Scientific 900062 Optimum Results B ISE Fill Solution; Cl/CN/Pb/ Ag/S
Thermo Scientific | Mfr: 900062| Item: 0571332
Filling solutions for use with pH electrodes and reference electrodes
₹30,700.00INR / Each
₹30,700.00INR / Each
Buy It Together
Current Item
Thermo Scientific 900062 Optimum Results B ISE Fill Solution; Cl/CN/Pb/ Ag/S
Thermo Scientific 900062 Optimum Results B ISE Fill Solution; Cl/CN/Pb/ Ag/S
0571332
₹30,700.00INR / Each
+ +
Thermo Scientific 810001 Storage Soluton, 475 mL bottle
0573060
₹18,000.00INR / Each
+ =
Total Price 145710.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Solutions work best when combined with ionic strength and pH adjusters, which eliminate interferences from other ion species
  • 300 mL total: 5 x 60 mL bottles
More About this Item
All filling solutions for reference electrodes come in cartons of one 60-mL or five 50-mL bottles.
Specifications
  • Ion-Selective ElectrodeChloride, Cyanide, Lead, Silver/Sulfide
  • DescriptionOptimum Results B ISE Fill Solution; Cl/CN/Pb/ Ag/S