Thermo Scientific 900017 Filling solutions: use with Cl- electrodes
Representative image only.
Thermo Scientific 900017 Filling solutions: use with Cl- electrodes
Thermo Scientific | Mfr: 900017| Item: 0573047
Filling solutions for use with pH electrodes and reference electrodes
₹50,100.00INR / Case of 5
₹50,100.00INR / Case of 5
Buy It Together
Current Item
Thermo Scientific 900017 Filling solutions: use with Cl- electrodes
Thermo Scientific 900017 Filling solutions: use with Cl- electrodes
0573047
₹50,100.00INR / Case of 5
+ +
Thermo Scientific 810001 Storage Soluton, 475 mL bottle
0573060
₹18,000.00INR / Each
+ =
Total Price 165110.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Solutions work best when combined with ionic strength and pH adjusters, which eliminate interferences from other ion species
More About this Item
All filling solutions for reference electrodes come in cartons of one 60-mL or five 50-mL bottles.
Specifications
  • Ion-Selective ElectrodeChloride
  • DescriptionFilling solutions: use with Cl- electrodes