Thermo Scientific 8157BNURCA ROSS Ultra pH/ATC Probe, DJ/Epoxy, NTC30/RCA
Representative image only.
Thermo Scientific 8157BNURCA ROSS Ultra pH/ATC Probe, DJ/Epoxy, NTC30/RCA
Thermo Scientific | Mfr: 8157BNURCA| Item: 0571808
18 months replacement warranty
₹142,000.00INR / Each
₹142,000.00INR / Each
Buy It Together
Current Item
Thermo Scientific 8157BNURCA ROSS Ultra pH/ATC Probe, DJ/Epoxy, NTC30/RCA
Thermo Scientific 8157BNURCA ROSS Ultra pH/ATC Probe, DJ/Epoxy, NTC30/RCA
0571808
₹142,000.00INR / Each
+ + +
Thermo Scientific 810007 electrode fill solution, 3 M KCl 5/cs
0573052
₹24,600.00INR / Case of 5
=
Total Price 223500.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Builds on the ROSS® technology for greater stability, relability, and renowned accuracy and temperature response
  • Less long term drift saves time from recalibrating
  • Double junction
  • Epoxy body
  • NTC30 temperature compensation
  • RCA connetion
  • 2 year warranty
More About this Item
The ROSS Ultra stability gives you ±0.01 pH accuracy even over a long period of time. Using a nonmetallic reference system, the liquid-liquid equilibration occurs almost instantaneously, saving time plus ensuring accurate results. Complex, delicate samples, such as biological media, foods, and pharmaceuticals, measure easily without metallic contamination of the sample or excessive cleaning. ROSS Ultra electrodes are refillable—easily accommodating a change in filling solution for samples sensitive to potassium or chloride ions. Choose ROSS Ultra pH/ATC Triodes for applications requiring built-in temperature compensation.
For use with Mettler Five and Seven Series meters.
Two year warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialEpoxy
  • Reference JunctionGlass fiber
  • Reference ElectrolyteROSS
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature CompensationYes
  • ConnectorNTC30 with RCA
  • Cable Length (m)1
  • DescriptionUltra pH/ATC Probe, DJ/Epoxy, NTC30/RCA