Thermo Scientific 8107UWMMD ROSS Ultra pH/ATC Probe, DJ/Epoxy/3m; BNC/8-Pin DIN
Representative image only.
Thermo Scientific 8107UWMMD ROSS Ultra pH/ATC Probe, DJ/Epoxy/3m; BNC/8-Pin DIN
Thermo Scientific | Mfr: 8107UWMMD| Item: 0571848
18 months replacement warranty
₹207,000.00INR / Each
₹207,000.00INR / Each
Buy It Together
Current Item
Thermo Scientific 8107UWMMD ROSS Ultra pH/ATC Probe, DJ/Epoxy/3m; BNC/8-Pin DIN
Thermo Scientific 8107UWMMD ROSS Ultra pH/ATC Probe, DJ/Epoxy/3m; BNC/8-Pin DIN
0571848
₹207,000.00INR / Each
+ + =
Total Price 263900.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Builds on the ROSS® technology for greater stability, relability, and renowned accuracy and temperature response
  • Less long term drift saves time from recalibrating
  • Double junction
  • Epoxy body
  • 8-Pin DIN temperature compensation
  • BNC connection
  • 3 m cable length
  • 18 month warranty
More About this Item
The ROSS Ultra stability gives you ±0.01 pH accuracy even over a long period of time. Using a nonmetallic reference system, the liquid-liquid equilibration occurs almost instantaneously, saving time plus ensuring accurate results. Complex, delicate samples, such as biological media, foods, and pharmaceuticals, measure easily without metallic contamination of the sample or excessive cleaning. ROSS Ultra electrodes are refillable—easily accommodating a change in filling solution for samples sensitive to potassium or chloride ions. Choose ROSS Ultra pH/ATC Triodes for applications requiring built-in temperature compensation.
18 Month Warranty
Specifications
  • ApplicationsSealed, gel-filled
  • Electrode TypeDouble junction
  • Electrode Body MaterialEpoxy
  • Reference JunctionGlass capillary
  • Reference ElectrolyteROSS
  • Min pH (pH)0
  • Max pH (pH)14
  • Temperature CompensationYes
  • Connector8-pin mini-DIN
  • Cable Length (m)3
  • DescriptionUltra pH/ATC Probe, DJ/Epoxy/3m; BNC/8-Pin DIN
  • Warranty18 months