Thermo Scientific 8102BNUWP ROSS Ultra pH Probe, Refillable/DJ/Glass; WP BNC
Representative image only.
Thermo Scientific 8102BNUWP ROSS Ultra pH Probe, Refillable/DJ/Glass; WP BNC
Thermo Scientific | Mfr: 8102BNUWP| Item: 0571870
18 months replacement warranty
₹80,440.00INR / Each
₹80,440.00INR / Each
Buy It Together
Current Item
Thermo Scientific 8102BNUWP ROSS Ultra pH Probe, Refillable/DJ/Glass; WP BNC
Thermo Scientific 8102BNUWP ROSS Ultra pH Probe, Refillable/DJ/Glass; WP BNC
0571870
₹80,440.00INR / Each
+ + =
Total Price 137340.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Builds on the ROSS® technology for greater stability, relability, and renowned accuracy and temperature response
  • Less long term drift saves time from recalibrating
  • 2 year warranty
More About this Item
The ROSS Ultra stability gives you ±0.01 pH accuracy even over a long period of time. Using a nonmetallic reference system, the liquid-liquid equilibration occurs almost instantaneously, saving time plus ensuring accurate results. Complex, delicate samples, such as biological media, foods, and pharmaceuticals, measure easily without metallic contamination of the sample or excessive cleaning. ROSS Ultra electrodes are refillable—easily accommodating a change in filling solution for samples sensitive to potassium or chloride ions. Choose ROSS Ultra pH/ATC Triodes for applications requiring built-in temperature compensation.
Two year warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference JunctionGlass fiber
  • Reference ElectrolyteROSS
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature CompensationNo
  • ConnectorBNC
  • Cable Length (m)1
  • DescriptionUltra pH Probe, Refillable/DJ/Glass; WP BNC