Thermo Scientific 810199 All-in-One pH Buffer Kit with Cleaning Solution
Representative image only.
Thermo Scientific 810199 All-in-One pH Buffer Kit with Cleaning Solution
Thermo Scientific | Mfr: 810199| Item: 5535066
Common calibration values are color-coded for ease of use
Call For Price
Call For Price
Buy It Together
Current Item
Thermo Scientific 810199 All-in-One pH Buffer Kit with Cleaning Solution
Thermo Scientific 810199 All-in-One pH Buffer Kit with Cleaning Solution
5535066
Call For Price
+ =
Total Price 3600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
475 mL bottles of each 4.01, 7.00, and 10.01 pH buffers, as well as pH electrode storage solution, and an electrode storage bottle. And additional 60-mL bottle of pH electrode storage solution is included as well.
Key Features
  • Traceable to NIST standards
  • 475 mL bottles of pH 4.01, 7.00 and 10.01 buffers
More About this Item
Each 475-mL (pint) buffer bottle comes color-coded for easy identification and packaged with buffer/temperature tables for accurate calibrations. Buffers are standardized against NIST-traceable pH references for superior accuracy.
NIST Traceable Calibration
Specifications
  • Buffer Kit Values4.01, 7.00, 10.01 pH buffers, storage solution, cleaning solution, and storage bottle
  • Volume (mL)475
  • ColorColored
  • DescriptionAll-in-One pH Buffer Kit with Cleaning Solution