Thermo Scientific 810017 pH Electrode Storage Sleeve
Representative image only.
Thermo Scientific 810017 pH Electrode Storage Sleeve
Thermo Scientific | Mfr: 810017| Item: 9957050
Designed to condition and protect up to 12-mm dia
₹21,900.00INR / Each
₹21,900.00INR / Each
Product Description
Key Features
  • Protect fragile pH glass bulbs from breakage
  • Keep electrodes conditioned and ready for pH measurement
  • Weighted base allows for safe, upright storage
More About this Item
Storage sleeve and base help simplify the storage of your electrodes. Made from durable polypropylene, the sleeve accommodates all electrodes with a 12-mm diameter and up to a 120-mm length. The sleeve is easily removed to use electrode for measurement.
Specifications
  • MaterialPolypropylene (PP)
  • Compatible WithAll brands of electrodes
  • DescriptionpH Electrode Storage Sleeve