Thermo Scientific 5520 Sterile Electroporation Cuvette, 2 mm gap width, blue cap, 50/pack
Representative image only.
Thermo Scientific 5520 Sterile Electroporation Cuvette, 2 mm gap width, blue cap, 50/pack
Thermo Scientific | Mfr: 5520| Item: 2571402
High-quality disposable cuvettes are thoroughly cleansed to eliminate trace inhibitor materials
₹102,000.00INR / Pkg of 50
₹102,000.00INR / Pkg of 50
Product Description
Includes
presterilized transfer pipette.
Key Features
  • Limited Lifetime Warranty
More About this Item
Each cuvette is sterile and individually packed with a presterilized transfer pipette. Color coded frosted caps enable trouble-free labeling and recognition. Sturdy polycarbonate construction withstands pulses of very high voltages or field strength and is autoclavable. Compatible with most electroporation systems.
These cuvettes have a 2mm cap width which allows fo rmedium field strength- ideal for yeast cells.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Limited Lifetime WarrantySterile
Specifications
  • MaterialPolycarbonate
  • DescriptionSterile Electroporation Cuvette, 2 mm gap width, blue cap, 50/pack
  • WarrantyLimited Lifetime