Thermo Scientific 5510-11 Sterile Electroporation Cuvette, 1 mm gap width, white cap, 50/pack
Representative image only.
Thermo Scientific 5510-11 Sterile Electroporation Cuvette, 1 mm gap width, white cap, 50/pack
Thermo Scientific | Mfr: 5510-11| Item: 2571400
High-quality disposable cuvettes are thoroughly cleansed to eliminate trace inhibitor materials
₹102,000.00INR / Pkg of 50
₹102,000.00INR / Pkg of 50
Product Description
Includes
presterilized transfer pipette.
Key Features
  • Limited Lifetime Warranty
More About this Item
Each cuvette is sterile and individually packed with a presterilized transfer pipette. Color coded frosted caps enable trouble-free labeling and recognition. Sturdy polycarbonate construction withstands pulses of very high voltages or field strength and is autoclavable. Compatible with most electroporation systems.
These electroporation pipettes have a 1mm gap width which allows for highest field of strength- ideal for bacterial cells.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Limited Lifetime WarrantySterile
Specifications
  • MaterialPolycarbonate
  • DescriptionSterile Electroporation Cuvette, 1 mm gap width, white cap, 50/pack
  • WarrantyLimited Lifetime