Thermo Scientific 300349 Adhesive Flask Mat, 9" x 9"; 1 Roll
Representative image only.
Thermo Scientific 300349 Adhesive Flask Mat, 9" x 9"; 1 Roll
Thermo Scientific | Mfr: 300349| Item: 5170945
Enhance your platform shakers
₹24,300.00INR / Each
₹24,300.00INR / Each
Buy It Together
Current Item
Thermo Scientific 300349 Adhesive Flask Mat, 9" x 9"; 1 Roll
Thermo Scientific 300349 Adhesive Flask Mat, 9" x 9"; 1 Roll
5170945
₹24,300.00INR / Each
+ =
Total Price 136300.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 3 month warranty
More About this Item
Choose from multiple universal accessory options to expand your Thermo Scientific platform shakers.
The adhesive flask mat is an alternative to flask clamps for securing odd shaped objects such as bottles, volumetric flasks, and test tube racks to a shaker platform. Simply place the mat on an anodized aluminum or stainless steel shaker platform and secure the vessel to it.
3 Month Warranty
Specifications
  • DescriptionAdhesive Flask Mat (1 roll) 9" x 9"