Thermo Scientific 160-0040 Bath Lid; Flat with Handle
Representative image only.
Thermo Scientific 160-0040 Bath Lid; Flat with Handle
Thermo Scientific | Mfr: 1600040| Item: 1214426
Ideal for optimal use of your circulating bath
₹58,300.00INR / Each
₹58,300.00INR / Each
Buy It Together
Current Item
Thermo Scientific 160-0040 Bath Lid; Flat with Handle
Thermo Scientific 160-0040 Bath Lid; Flat with Handle
1214426
₹58,300.00INR / Each
+ =
Total Price 58300.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 3 year warranty
More About this Item
These stainless steel bath covers ensure temperatures are maintained in your circulating water baths.
For 29 to 51-Liter models 12144-20, -25, -50, -55, -85; 12147-20, -25, -50, -55.
Conformite Europeenne3 Year Warranty
Specifications
  • DescriptionBath Lid; Flat with Handle